accessibility menu, dialog, popup

UserName

H3COOCl

CAS RN 20395-28-2

IUPAC Name: 5-chloropentyl acetate
Synonyms: 5-chloropentyl acetate
Molecular Weight (g/mol): 164.63
Molecular Formula: C7H13ClO2
InChi Key: ZCYVIAZIVJNAMO-UHFFFAOYSA-N
SMILES: CC(=O)OCCCCCCl
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (1)
  • (1)