missing translation for 'onlineSavingsMsg'
Learn More

5-Chloroindole-2-carboxylic acid, 98%

Product Code. 11468567
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11468567 25 g 25g
11448567 1 g 1g
11458567 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11468567 Supplier Thermo Scientific Alfa Aesar Supplier No. A18626.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 10517-21-2 | C9H5ClNO2 | 194.59 g/mol

Potent antagonist at the glycine site of the NMDA receptor

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10517-21-2
Molecular Formula C9H5ClNO2
Molecular Weight (g/mol) 194.59
MDL Number MFCD00005613
InChI Key FUQOTYRCMBZFOL-UHFFFAOYSA-M
Synonym 5-chloroindole-2-carboxylic acid, 5-chloroindole-2-carboxylate, 1h-indole-2-carboxylic acid, 5-chloro, spectrum_000183, 5-chloroindoline-2-carboxylicacid, pubchem1670, acmc-1bztj, specplus_000670, spectrum2_000513, spectrum3_000783
PubChem CID 82693
SMILES [O-]C(=O)C1=CC2=CC(Cl)=CC=C2N1

Specifications

Melting Point 287°C (decomposition)
Quantity 25 g
Beilstein 153229
IUPAC Name 5-chloro-1H-indole-2-carboxylic acid
Formula Weight 195.6
Percent Purity 98%
Chemical Name or Material 5-Chloroindole-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.