missing translation for 'onlineSavingsMsg'
Learn More

5-(bromomethyl)-3-methylisoxazole, 95%, Thermo Scientific™

Product Code. 10208563
Change view
Click to view available options
Quantity:
250 mg
Packaging:
Amber Glass Bottle
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10208563 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10208563 Supplier Thermo Scientific Maybridge Supplier No. CC27008CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 36958-61-9
Molecular Formula C5H6BrNO
Molecular Weight (g/mol) 176.013
MDL Number MFCD09879901
InChI Key DMUJLHLWCUMHRT-UHFFFAOYSA-N
Synonym 5-bromomethyl-3-methylisoxazole, 5-bromomethyl-3-methyl-1,2-oxazole, isoxazole, 5-bromomethyl-3-methyl, 3-methyl-5-bromomethylisoxazole
PubChem CID 10607354
IUPAC Name 5-(bromomethyl)-3-methyl-1,2-oxazole
SMILES CC1=NOC(=C1)CBr

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.01
Percent Purity 95%
Chemical Name or Material 5-(Bromomethyl)-3-methylisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.