accessibility menu, dialog, popup

UserName

CH3NOBr

CAS RN 36958-61-9

IUPAC Name: 5-(bromomethyl)-3-methyl-1,2-oxazole
Synonyms: 5-(bromomethyl)-3-methyl-1,2-oxazole
Molecular Weight (g/mol): 176.01
Molecular Formula: C5H6BrNO
InChi Key: DMUJLHLWCUMHRT-UHFFFAOYSA-N
SMILES: CC1=NOC(CBr)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results