missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-N,N-dimethylpyridine-2-carboxamide, 96%, Thermo Scientific™

Product Code. 15490898
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490898 1 g 1g
15441486 250 mg 250mg
15400908 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490898 Supplier Thermo Scientific Alfa Aesar Supplier No. H54686.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
5-Bromo-N,N-dimethylpyridine-2-carboxamide is used as an organic chemical synthesis intermediate.

Notes
Air sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 845305-86-4
Molecular Formula C8H9BrN2O
Molecular Weight (g/mol) 229.077
MDL Number MFCD07357419
InChI Key KQRHFAPOFNDZHY-UHFFFAOYSA-N
Synonym 5-bromo-n,n-dimethylpicolinamide, 5-bromopyridine-2-carboxylic acid dimethylamide, 5-bromo-pyridine-2-carboxylic acid dimethylamide, 5-bromo-n,n-dimethyl-pyridine-2-carboxamide
PubChem CID 22831618
IUPAC Name 5-bromo-N,N-dimethylpyridine-2-carboxamide
SMILES CN(C)C(=O)C1=NC=C(C=C1)Br

Specifications

Quantity 1 g
Formula Weight 229.07
Percent Purity 96%
Chemical Name or Material 5-Bromo-N,N-dimethylpyridine-2-carboxamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.