accessibility menu, dialog, popup

UserName

CH3NH3CONBr

CAS RN 845305-86-4

IUPAC Name: 5-bromo-N,N-dimethylpyridine-2-carboxamide
Synonyms: 5-bromo-N,N-dimethylpyridine-2-carboxamide
Molecular Weight (g/mol): 229.08
Molecular Formula: C8H9BrN2O
InChi Key: KQRHFAPOFNDZHY-UHFFFAOYSA-N
SMILES: CN(C)C(=O)C1=NC=C(Br)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results