missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-methoxycinnamic acid, 98%, Thermo Scientific™

Catalog Number 11433388
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
40803-53-0
C10H8BrO3
256.08
MFCD00017596
QABZTGNRWBRJBO-GORDUTHDSA-M
5-bromo-2-methoxycinnamic acid, 3-5-bromo-2-methoxyphenyl acrylic acid, 2e-3-5-bromo-2-methoxyphenyl prop-2-enoic acid, 2e-3-5-bromo-2-methoxyphenyl acrylic acid, e-3-5-bromo-2-methoxyphenyl acrylic acid, substance h 47, 3-bromo-6-methoxycinnamic acid, acide bromo-5-methoxy-2-cinnamique, trans-3-bromo-6-methoxycinnamic acid, 3-5-bromo-2-methoxyphenyl propenoic acid
676162
(E)-3-(5-bromo-2-methoxyphenyl)prop-2-enoic acid
COC1=CC=C(Br)C=C1\C=C\C([O-])=O
This item is not returnable. View return policy
40803-53-0
C10H8BrO3
256.08
MFCD00017596
QABZTGNRWBRJBO-GORDUTHDSA-M
5-bromo-2-methoxycinnamic acid, 3-5-bromo-2-methoxyphenyl acrylic acid, 2e-3-5-bromo-2-methoxyphenyl prop-2-enoic acid, 2e-3-5-bromo-2-methoxyphenyl acrylic acid, e-3-5-bromo-2-methoxyphenyl acrylic acid, substance h 47, 3-bromo-6-methoxycinnamic acid, acide bromo-5-methoxy-2-cinnamique, trans-3-bromo-6-methoxycinnamic acid, 3-5-bromo-2-methoxyphenyl propenoic acid
676162
(E)-3-(5-bromo-2-methoxyphenyl)prop-2-enoic acid
COC1=CC=C(Br)C=C1\C=C\C([O-])=O

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 224°C to 226°C
Quantity 5 g
Beilstein 3265594
Formula Weight 257.09
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-methoxycinnamic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'