missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-iodobenzoic acid, 97%

Product Code. 15421929
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15421929 5 g 5g
15431929 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15421929 Supplier Thermo Scientific Alfa Aesar Supplier No. H64643.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21740-00-1 | C7H4BrIO2 | 326.915 g/mol

It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis agrochemical, pharmaceutical and dyestuff field.

Solubility
Slightly soluble in water.

Notes
Light Sensitive. Store in dark. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21740-00-1
Molecular Formula C7H4BrIO2
Molecular Weight (g/mol) 326.915
MDL Number MFCD00079717
InChI Key IGBNDUKRHPTOBP-UHFFFAOYSA-N
Synonym 2-iodo-5-bromobenzoic acid, 5-bromo-2-iodo-benzoic acid, benzoic acid, 5-bromo-2-iodo, 5-bromo-2-iodobenzoicacid, pubchem3789, acmc-1bfz2, ksc497i6j, rarechem al bo 0892, akos bbv-001713, 5-bromo-2-iodobenzoic acid
PubChem CID 302939
IUPAC Name 5-bromo-2-iodobenzoic acid
SMILES C1=CC(=C(C=C1Br)C(=O)O)I

Specifications

Melting Point 158°C to 163°C
Quantity 5 g
Beilstein 3247380
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 326.92
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Bromo-2-iodobenzoic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.