accessibility menu, dialog, popup

UserName

OOBrI

CAS RN 21740-00-1

IUPAC Name: 5-bromo-2-iodobenzoate
Synonyms: 5-bromo-2-iodobenzoate
Molecular Weight (g/mol): 325.91
Molecular Formula: C7H3BrIO2
InChi Key: IGBNDUKRHPTOBP-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC(Br)=CC=C1I

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results