missing translation for 'onlineSavingsMsg'
Learn More

alpha-Cyano-4-hydroxycinnamic acid, Ultrapure MALDI Matrix, Thermo Scientific Chemicals

Product Code. 15410777
Change view
Click to view available options
Quantity:
5 x 10 mg
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15410777 5 x 10 mg Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15410777 Supplier Thermo Scientific Alfa Aesar Supplier No. J67635.!K
 Restricted Item
Estimated Shipment: 16-09-2026
Add to basket
This item is not returnable. View return policy

 

For use as a matrix in MALDI mass spectrometry analysis

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 28166-41-8
Melting Point 250°C to 253°C
Color Yellow
Odor Odorless
Molecular Formula C10H7NO3
MDL Number MFCD00004204
Quantity 5 x 10 mg
Beilstein 3271427
Synonym alpha-cyano-4-hydroxycinnamate, 2-cyano-4-hydroxycinnamate, 2-cyano-3-4-hydroxyphe-nyl acrylic acid, 2z-2-cyano-3-4-hydroxyphenyl prop-2-enoic acid
InChI Key AFVLVVWMAFSXCK-YVMONPNESA-N
SMILES C1=CC(=CC=C1C=C(C#N)C(=O)O)O
IUPAC Name (Z)-2-cyano-3-(4-hydroxyphenyl)prop-2-enoic acid
Molecular Weight (g/mol) 189.17
PubChem CID 9794103
Formula Weight 189.17
Physical Form Powder
Chemical Name or Material alpha-Cyano-4-hydroxycinnamic acid, Ultrapure MALDI Matrix
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.