missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-fluorocinnamic acid, 98%, Thermo Scientific™

Product Code. 11459200
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459200 5 g 5g
11469200 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11459200 Supplier Thermo Scientific Alfa Aesar Supplier No. A16109.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 202865-71-2
Molecular Formula C9H6BrFO2
Molecular Weight (g/mol) 245.047
MDL Number MFCD00143427
InChI Key ROTBALNVXDGIQY-DAFODLJHSA-N
Synonym 5-bromo-2-fluorocinnamic acid, 2e-3-5-bromo-2-fluorophenyl prop-2-enoic acid, 3-5-bromo-2-fluorophenyl prop-2-enoic acid, 3-5-bromo-2-fluoro-phenyl-acrylic acid, e-3-5-bromo-2-fluorophenyl acrylic acid, 3-5-bromo-2-fluorophenyl-2-propenoic acid, pubchem3230, rarechem bk hw 0189, timtec-bb sbb006748, attercop-chm at116397
PubChem CID 688316
IUPAC Name (E)-3-(5-bromo-2-fluorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1Br)C=CC(=O)O)F

Specifications

Melting Point 193°C to 196°C
Quantity 5 g
Formula Weight 245.05
Percent Purity 98%
Chemical Name or Material 5-Bromo-2-fluorocinnamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.