missing translation for 'onlineSavingsMsg'
Learn More

5-Bromo-2-ethoxybenzonitrile, 99%, Thermo Scientific™

Product Code. 11430140
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11430140 250 mg 250mg
11440140 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430140 Supplier Thermo Scientific Alfa Aesar Supplier No. H50624.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 279263-03-5
Molecular Formula C9H8BrNO
Molecular Weight (g/mol) 226.07
MDL Number MFCD02257397
InChI Key RNRDIQJUDTUNEN-UHFFFAOYSA-N
PubChem CID 1228878
IUPAC Name 5-bromo-2-ethoxybenzonitrile
SMILES CCOC1=C(C=C(Br)C=C1)C#N

Specifications

Melting Point 82°C to 84°C
Quantity 250 mg
UN Number UN3439
Synonym 5-bromo-2-ethoxy-benzonitrile, benzonitrile, 5-bromo-2-ethoxy, 5-bromo-2-ethoxybenzenecarbonitrile, acmc-1cksx, benzonitrile,5-bromo-2-ethoxy
Formula Weight 226.07
Percent Purity 99%
Chemical Name or Material 5-Bromo-2-ethoxybenzonitrile
compliance-icons
Product Identifier
  • 5-Bromo-2-ethoxybenzonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: 5-bromo-2-ethoxybenzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.