accessibility menu, dialog, popup

UserName

CAS RN 279263-03-5

CH3OBrN

CAS RN 279263-03-5

IUPAC Name: 5-bromo-2-ethoxybenzonitrile
Synonyms: 5-bromo-2-ethoxybenzonitrile
Molecular Weight (g/mol): 226.07
Molecular Formula: C9H8BrNO
InChi Key: RNRDIQJUDTUNEN-UHFFFAOYSA-N
SMILES: CCOC1=C(C=C(Br)C=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results