missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Aminoindole hydrochloride, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Specifications
CAS Min % | 94.0 |
CAS Max % | 100.0 |
Melting Point | 253.0°C |
Color | Brown to Tan |
Infrared Spectrum | Authentic |
Assay Percent Range | 94% min. (HPLC) |
Packaging | Glass bottle |
Quantity | 250 mg |
Formula Weight | 168.63 |
Percent Purity | 95% |
Show More |
Safety and Handling
Product Identifier
- 5-Aminoindole hydrochloride
- Warning
- Acute toxicity Category 4
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'