accessibility menu, dialog, popup

UserName

HNNH2HCl

CAS RN 65795-92-8

IUPAC Name: 1H-indol-5-amine hydrochloride
Synonyms: 5-aminoindole hydrochloride
Molecular Weight (g/mol): 168.62
Molecular Formula: C8H9ClN2
InChi Key: CZRIFWIECZRIKN-UHFFFAOYSA-N
SMILES: Cl.NC1=CC=C2NC=CC2=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results