missing translation for 'onlineSavingsMsg'
Learn More

5-Aminoindole hydrochloride, 95%, Thermo Scientific™

Product Code. 10041084 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041084 1 g Glass bottle 1g
10224784 250 mg Glass bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041084 Supplier Thermo Scientific Acros Supplier No. 426620010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 65795-92-8
Molecular Formula C8H8N2·HCl
Molecular Weight (g/mol) 168.63
MDL Number MFCD00077647
InChI Key CZRIFWIECZRIKN-UHFFFAOYSA-N
Synonym 5-aminoindole hydrochloride, 1h-indol-5-amine hydrochloride, 5-aminoindole monohydrochloride, 5-aminoindole hcl, 5-aminoindol-hydrochloride, 5-indolamine hydrochloride, 1h-indol-5-amine,hydrochloride
PubChem CID 16211959
IUPAC Name 1H-indol-5-amine;hydrochloride
SMILES C1=CC2=C(C=CN2)C=C1N.Cl

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 253.0°C
Color Brown to Tan
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 168.63
Percent Purity 95%
Physical Form Crystals
Chemical Name or Material 5-Aminoindole hydrochloride
Show More Show Less
compliance-icons
Product Identifier
  • 5-Aminoindole hydrochloride
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.