missing translation for 'onlineSavingsMsg'
Learn More

5-Amino-2-(4-morpholinyl)pyridine, 97%

Product Code. 15467887
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15467887 5 g 5g
11490250 250 mg 250mg
11400260 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15467887 Supplier Thermo Scientific Alfa Aesar Supplier No. H50699.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 52023-68-4 | C9H13N3O | 179.223 g/mol

5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Amino-2-(4-morpholinyl)pyridine is used as pharmaceutical intermediate.

Notes
Store at room temperature. Store in a cool, dry condition in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52023-68-4
Molecular Formula C9H13N3O
Molecular Weight (g/mol) 179.223
MDL Number MFCD00119562
InChI Key VVTSPTCBHTWXMD-UHFFFAOYSA-N
Synonym 6-morpholinopyridin-3-amine, 3-amino-6-morpholinopyridine, 6-morpholino-3-pyridinamine, 6-morpholin-4-yl pyridin-3-amine, 6-morpholino pyridin-3-amine, 6-morpholinopyridine-3-amine, 3-pyridinamine, 6-4-morpholinyl, 6-morpholin-4-yl-3-pyridylamine, 5-amino-2-morpholin-4-ylpyridine, 5-amino-2-4-morpholinyl pyridine
PubChem CID 104059
IUPAC Name 6-morpholin-4-ylpyridin-3-amine
SMILES C1COCCN1C2=NC=C(C=C2)N

Specifications

Melting Point 127°C to 130°C
Quantity 5 g
Formula Weight 179.22
Percent Purity 97%
Chemical Name or Material 5-Amino-2-(4-morpholinyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.