accessibility menu, dialog, popup

UserName

CAS RN 52023-68-4

NH2HNNO

CAS RN 52023-68-4

IUPAC Name: 5-amino-2-(morpholin-4-yl)pyridin-1-ium
Synonyms: 5-amino-2-(morpholin-4-yl)pyridin-1-ium
Molecular Weight (g/mol): 180.23
Molecular Formula: C9H14N3O
InChi Key: VVTSPTCBHTWXMD-UHFFFAOYSA-O
SMILES: NC1=CC=C([NH+]=C1)N1CCOCC1
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (2)
  • (1)
  • (1)