missing translation for 'onlineSavingsMsg'
Learn More

5,7-Dichloro-4-hydroxyquinoline, TRC

Product Code. 30383739
Change view
Click to view available options
Quantity:
50 mg
250 mg
1 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30383739 250 mg
30377917 50 mg
30376214 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30383739 Supplier Toronto Research Chemicals Supplier No. TRCD437240250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 171850-29-6
Molecular Formula C9 H5 Cl2 N O
Molecular Weight (g/mol) 214.05
Synonym 5,7-Dichloro-4-quinolinol, 5,7-Dichloro-4-hydroxyquinoline
IUPAC Name 5,7-dichloroquinolin-4-ol
SMILES Oc1ccnc2cc(Cl)cc(Cl)c12

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C9H5Cl2NO/c10-5-3-6(11)9-7(4-5)12-2-1-8(9)13/h1-4H,(H,12,13)
Formula Weight 212.9748
Chemical Name or Material 5,7-Dichloro-4-hydroxyquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.