CAS RN 171850-29-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5,7-Dichloro-4-hydroxyquinoline, TRC
CAS: 171850-29-6 Molecular Formula: C9 H5 Cl2 N O Molecular Weight (g/mol): 214.05 Synonym: 5,7-Dichloro-4-quinolinol,5,7-Dichloro-4-hydroxyquinoline IUPAC Name: 5,7-dichloroquinolin-4-ol SMILES: Oc1ccnc2cc(Cl)cc(Cl)c12