missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ 5(6)-TAMRA (5-(and-6)-Carboxytetramethylrhodamine), mixed isomers
Description
5(6)-TAMRA contains a carboxylic acid that can be used to react with primary amines via carbodiimide activation of the carboxylic acid. This bright, orange-fluorescent dye produces conjugates with absorption/emission maxima of ∼555/580 nm.
Specifications
Specifications
| Quantity | 100 mg |
| Product Type | 5(6)-TAMRA |
| Content And Storage | Store at room temperature and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Carboxylic Acid |
| Label or Dye | TAMRA™ Isomers, TMR (Tetramethylrhodamine) |
| Label Type | Classic Dyes |
| Emission | 580 |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?