missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5(6)-FITC

Product Code. 16481796
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16481796 500 mg 500mg
16407006 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16481796 Supplier MedChemExpress Supplier No. HY15941
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5(6)-FITC (Fluorescein 5(6)-isothiocyanate) is an amine-reactive derivative of fluorescein dye that has wide-ranging applications as a label for antibodies and other probes, FC and IF-based assays such as WB and ELISA.

Specifications

Quantity 500 mg
Molecular Weight (g/mol) 389.38 g/mol
Solubility DMSO : ≥ 156 mg/mL (400.64 mM)
Content And Storage 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
CAS 27072-45-3
Synonym Fluorescein 5(6)-isothiocyanate ∣ Fluorescein isothiocyanate 5- and 6- isomers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.