missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ 5(6)-CR 6G, SE (5-(and-6)-Carboxyrhodamine 6G, Succinimidyl Ester), mixed isomers
Description
The excitation and emission spectra of carboxyrhodamine 6G (CR 6G) fall between those of fluorescein and tetramethylrhodamine. With a peak absorption at 525 nm, conjugates of carboxyrhodamine 6G are an excellent match to the 514 nm spectral line of the argon-ion laser. The amine-reactive 5(6)CR 6G, SE can be used to attach this fluorophore to biomolecules of interest.
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | 5(6)-CR 6G SE |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | Rhodamine 6G |
| Label Type | Classic Dyes |
| Emission | 525 |
| Excitation | 514 |
For Research Use Only. Not for use in diagnostic procedures.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?