missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress 5(6)-Carboxyfluorescein

Product Code. 16451589
Encompass_Preferred
Change view
Click to view available options
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
16451589 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16451589 Supplier MedChemExpress Supplier No. HY15940
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress 5(6)-Carboxyfluorescein (5(6)-FAM) is an amine-reactive pH-sensitive green fluorescent probe. 5(6)-Carboxyfluorescein (5(6)-FAM) can be used to label proteins, peptides and nucleotides.

5(6)-Carboxyfluorescein can be used for the detection of tumour areas in vivo.

Specifications

Quantity 1 g
Molecular Weight (g/mol) 752.63 g/mol
Solubility DMSO : 62.5 mg/mL (83.04 mM; ultrasonic and warming and heat to 60°C)
Content And Storage -20°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light)
Shipping Condition Room temperature
CAS 72088-94-9
Synonym 5(6)-FAM ∣ 5-(and-6)-Carboxyfluorescein mixed isomers
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.