missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MedChemExpress 5(6)-Aminofluorescein
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
Specifications
Specifications
| Quantity | 500 mg |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 347.32 g/mol |
| Solubility | DMSO : ≥ 39 mg/mL (112.29 mM) |
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Shipping Condition | Room temperature |
| Target | HBV |
| CAS | 27599-63-9 |
| Synonym | 5(6)-AFM ∣ Fluoresceinamine mixed isomers |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction