missing translation for 'onlineSavingsMsg'
Learn More

5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene, TRC

Product Code. 30418625
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418625 50 mg -
30421685 10 mg 10mg
30418904 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418625 Supplier Toronto Research Chemicals Supplier No. TRCT29474350MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53397-65-2
Molecular Formula C16H8
Molecular Weight (g/mol) 200.23
Synonym 5,6,11,12-Tetradehydrobenzo[a,e][8]annulene, Dibenzocyclooctadiyne, sym-Dibenzo-1,5-cyclooctadiene-3,7-diyne
IUPAC Name 5,6,11,12-tetradehydrodibenzo[2,1-a:2',1'-f][8]annulene
SMILES c1ccc2C#Cc3ccccc3C#Cc2c1

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C16H8/c1-2-6-14-11-12-16-8-4-3-7-15(16)10-9-13(14)5-1/h1-8H
Formula Weight 200.0626
Chemical Name or Material 5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.