CAS RN 53397-65-2
IUPAC Name:
tricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaen-2,10-diyne
Synonyms:
tricyclo[10.4.0.0⁴,⁹]hexadeca-1(16),4,6,8,12,14-hexaen-2,10-diyne
Molecular Weight (g/mol):
200.24
Molecular Formula:
C16H8
InChi Key:
JFYLRMKUYCIWMX-UHFFFAOYSA-N
SMILES:
C1=CC=C2C#CC3=CC=CC=C3C#CC2=C1
1
–
1
of
1
results
1
5,6,11,12-Tetradehydrodibenzo[a,e]cyclooctene, TRC
CAS: 53397-65-2 Molecular Formula: C16H8 Molecular Weight (g/mol): 200.23 Synonym: 5,6,11,12-Tetradehydrobenzo[a,e][8]annulene,Dibenzocyclooctadiyne,sym-Dibenzo-1,5-cyclooctadiene-3,7-diyne IUPAC Name: 5,6,11,12-tetradehydrodibenzo[2,1-a:2',1'-f][8]annulene SMILES: c1ccc2C#Cc3ccccc3C#Cc2c1