missing translation for 'onlineSavingsMsg'
Learn More

5-(4-Nitrophenyl)-2-furoic acid, Thermo Scientific™

Product Code. 10086452 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10086452 1 g 1g
10200113 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10086452 Supplier Thermo Scientific Acros Supplier No. 375530010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 28123-73-1
Molecular Formula C11H6NO5
Molecular Weight (g/mol) 232.17
MDL Number MFCD00094261
InChI Key FXZTYDPWMQBTDH-UHFFFAOYSA-M
Synonym 5-4-nitrophenyl furan-2-carboxylic acid, 5-4-nitrophenyl-2-furoic acid, 5-4-nitro-phenyl-furan-2-carboxylic acid, 2-furancarboxylic acid, 5-4-nitrophenyl, 2-furancarboxylic acid,5-4-nitrophenyl, 5-p-nitrophenyl-2-furancarboxylic acid, 5-4-nitrophenyl-2-furancarboxylic acid, 5-4-nitrophenyl-2-furan carboxylic acid
PubChem CID 161593
IUPAC Name 5-(4-nitrophenyl)furan-2-carboxylate
SMILES [O-]C(=O)C1=CC=C(O1)C1=CC=C(C=C1)[N+]([O-])=O

Specifications

Melting Point 255°C to 257°C
Packaging Glass bottle
Quantity 1 g
Formula Weight 233.18
Chemical Name or Material 5-(4-Nitrophenyl)-2-furoic acid
compliance-icons
Product Identifier
  • 5-(4-Nitrophenyl)-2-furoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
  • EUH044-Risk of explosion if heated under confinement.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.