accessibility menu, dialog, popup

UserName

OOONOO

CAS RN 28123-73-1

IUPAC Name: 5-(4-nitrophenyl)furan-2-carboxylate
Synonyms: 5-(4-nitrophenyl)furan-2-carboxylate
Molecular Weight (g/mol): 232.17
Molecular Formula: C11H6NO5
InChi Key: FXZTYDPWMQBTDH-UHFFFAOYSA-M
SMILES: [O-]C(=O)C1=CC=C(O1)C1=CC=C(C=C1)[N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results