missing translation for 'onlineSavingsMsg'
Learn More

5-[[4-(Hydroxymethyl)phenoxy]methyl]-3-(1-methylethyl)-2-oxazolidinone, TRC

Product Code. 30402683
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402683 50 mg 50mg
30394051 5 mg 5mg
30397459 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30402683 Supplier Toronto Research Chemicals Supplier No. TRCH82575550MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1071765-44-0
Molecular Formula C14 H19 N O4
Molecular Weight (g/mol) 265.3
Synonym 2-Oxazolidinone, 5-[[4-(hydroxymethyl)phenoxy]methyl]-3-(1-methylethyl)-, 5-[[4-(Hydroxymethyl)phenoxy]methyl]-3-isopropyl-1,3-oxazolidin-2-one, Bisoprolol Fumarate Imp. U (EP), Bisoprolol Oxazolidinonebenzylalcohol
IUPAC Name 5-[[4-(hydroxymethyl)phenoxy]methyl]-3-propan-2-yl-1,3-oxazolidin-2-one
SMILES CC(C)N1CC(COc2ccc(CO)cc2)OC1=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C14H19NO4/c1-10(2)15-7-13(19-14(15)17)9-18-12-5-3-11(8-16)4-6-12/h3-6,10,13,16H,7-9H2,1-2H3
Formula Weight 265.1314
Chemical Name or Material 5-[(4-(Hydroxymethyl)phenoxy)methyl]-3-isopropyl-1,3-oxazolidin-2-one (Bisoprolol Oxazolidinonebenzylalcohol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.