CAS RN 1071765-44-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
5-[[4-(Hydroxymethyl)phenoxy]methyl]-3-isopropyl-1,3-oxazolidin-2-one (Bisoprolol Oxazolidinonebenzylalcohol), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
5-[[4-(Hydroxymethyl)phenoxy]methyl]-3-(1-methylethyl)-2-oxazolidinone, TRC
CAS: 1071765-44-0 Molecular Formula: C14 H19 N O4 Molecular Weight (g/mol): 265.3 Synonym: 2-Oxazolidinone, 5-[[4-(hydroxymethyl)phenoxy]methyl]-3-(1-methylethyl)-,5-[[4-(Hydroxymethyl)phenoxy]methyl]-3-isopropyl-1,3-oxazolidin-2-one,Bisoprolol Fumarate Imp. U (EP),Bisoprolol Oxazolidinonebenzylalcohol IUPAC Name: 5-[[4-(hydroxymethyl)phenoxy]methyl]-3-propan-2-yl-1,3-oxazolidin-2-one SMILES: CC(C)N1CC(COc2ccc(CO)cc2)OC1=O