missing translation for 'onlineSavingsMsg'
Learn More

5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine, ≥97%, Thermo Scientific™

Product Code. 10515194
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10515194 1 g 1g
10301343 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10515194 Supplier Thermo Scientific Maybridge Supplier No. CC47839DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 250726-93-3
Molecular Formula C12H17BO4S
Molecular Weight (g/mol) 268.13
MDL Number MFCD08060541
InChI Key HRLHWIMNIQOHRF-UHFFFAOYSA-N
Synonym 2-2,3-dihydrothieno 3,4-b 1,4 dioxin-5-yl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-2,3-dihydrothieno 3,4-b 1,4 dioxine, 4,4,5,5-tetramethyl-2-2h,3h-thieno 3,4-b 1,4 dioxin-5-yl-1,3,2-dioxaborolane, 5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-2,3-dihydrothieno 3,4-b 1,4 dioxine, 2,3-dihydrothieno 3,4-b-1,4-dioxin-5-boronic acid pinacol ester, 2,3-dihydrothieno 3,4-b 1,4 dioxine-5-boronic acid pinacol ester, 2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl thieno 3,4-b 1,4 dioxin, thieno 3,4-b-1,4-dioxin,2,3-dihydro-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl
PubChem CID 15417854
IUPAC Name 5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine
SMILES CC1(C)OB(OC1(C)C)C1=C2OCCOC2=CS1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 1 g
Formula Weight 268.14
Percent Purity 95%
Chemical Name or Material 5-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-2,3-dihydrothieno[3,4-b][1,4]dioxine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.