accessibility menu, dialog, popup

UserName

BOOH3CH3CCH3H3CSOO

CAS RN 250726-93-3

IUPAC Name: 4,4,5,5-tetramethyl-2-{2H,3H-thieno[3,4-b][1,4]dioxin-5-yl}-1,3,2-dioxaborolane
Synonyms: 4,4,5,5-tetramethyl-2-{2H,3H-thieno[3,4-b][1,4]dioxin-5-yl}-1,3,2-dioxaborolane
Molecular Weight (g/mol): 268.13
Molecular Formula: C12H17BO4S
InChi Key: HRLHWIMNIQOHRF-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=C2OCCOC2=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results