missing translation for 'onlineSavingsMsg'
Learn More

5-(2-Phenyleth-1-ynyl)nicotinic acid, 97%, Thermo Scientific™

Product Code. 10279132
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10279132 250 mg Amber Glass Bottle 250mg
10762113 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10279132 Supplier Thermo Scientific Maybridge Supplier No. KM08328CB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175203-69-7
Molecular Formula C14H9NO2
Molecular Weight (g/mol) 223.231
MDL Number MFCD00174055
InChI Key DXJZBANECHYHRT-UHFFFAOYSA-N
Synonym 5-2-phenyleth-1-ynyl nicotinic acid, 5-2-phenylethynyl pyridine-3-carboxylic acid, 5-phenylethynyl pyridine-3-carboxylic acid, 3-pyridinecarboxylicacid, 5-2-phenylethynyl, maybridge1_004925, 5-phenylethynylnicotinic acid, 5-phenylethynyl-nicotinic acid, 5-phenylethynyl nicotinic acid, 5-phenylethynyl-pyridine-3-carboxylic acid
PubChem CID 2779236
IUPAC Name 5-(2-phenylethynyl)pyridine-3-carboxylic acid
SMILES C1=CC=C(C=C1)C#CC2=CC(=CN=C2)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 223.23
Percent Purity 97%
Chemical Name or Material 5-(2-Phenyleth-1-ynyl)nicotinic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.