accessibility menu, dialog, popup

UserName

NOOH

CAS RN 175203-69-7

IUPAC Name: 5-(2-phenylethynyl)pyridine-3-carboxylic acid
Synonyms: 5-(2-phenylethynyl)pyridine-3-carboxylic acid
Molecular Weight (g/mol): 223.23
Molecular Formula: C14H9NO2
InChi Key: DXJZBANECHYHRT-UHFFFAOYSA-N
SMILES: OC(=O)C1=CN=CC(=C1)C#CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results