missing translation for 'onlineSavingsMsg'
Learn More

5-(1-Piperazinyl)-2-pyridinamine, TRC

Product Code. 30418869
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30418869 100 mg 100mg
30411406 500 mg 500mg
30415619 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30418869 Supplier Toronto Research Chemicals Supplier No. TRCP482423100MG
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1082876-26-3
Molecular Formula C9H14N4
Molecular Weight (g/mol) 178.23
Synonym 5-(Piperazin-1-yl)pyridin-2-ylamine
IUPAC Name 5-piperazin-1-ylpyridin-2-amine
SMILES Nc1ccc(cn1)N2CCNCC2

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H14N4/c10-9-2-1-8(7-12-9)13-5-3-11-4-6-13/h1-2,7,11H,3-6H2,(H2,10,12)
Formula Weight 178.1218
Chemical Name or Material 5-(1-Piperazinyl)-2-pyridinamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.