CAS RN 1082876-26-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
5-(1-Piperazinyl)-2-pyridinamine, TRC
CAS: 1082876-26-3 Molecular Formula: C9H14N4 Molecular Weight (g/mol): 178.23 Synonym: 5-(Piperazin-1-yl)pyridin-2-ylamine IUPAC Name: 5-piperazin-1-ylpyridin-2-amine SMILES: Nc1ccc(cn1)N2CCNCC2