Learn More
5-(1,3-Oxazol-5-yl)-2-thiophenesulfonyl chloride, 97%, Thermo Scientific™
£138.00 - £401.00
Chemical Identifiers
CAS | 321309-40-4 |
---|---|
Molecular Formula | C7H4ClNO3S2 |
Molecular Weight (g/mol) | 249.68 |
MDL Number | MFCD02681961 |
InChI Key | MAGTZUMWQUDWAH-UHFFFAOYSA-N |
Synonym | 5-1,3-oxazol-5-yl-2-thiophenesulfonyl chloride, 5-1,3-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-oxazol-5-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-5-oxazolyl, 2-thiophenesulfonyl chloride, 5-5-oxazolyl, chloro 5-1,3-oxazol-5-yl 2-thienyl sulfone, 5-1,3-oxazol-5-yl thiophene-2-sulphonyl chloride, 5-1,3-oxazol-5-yl-2-thiophene sulfonyl chloride, 2-thiophenesulfonyl chloride, 5-5-oxazolyl-9ci |
PubChem CID | 2776411 |
IUPAC Name | 5-(1,3-oxazol-5-yl)thiophene-2-sulfonyl chloride |
SMILES | ClS(=O)(=O)C1=CC=C(S1)C1=CN=CO1 |
Description
Chemical Identifiers
321309-40-4 | |
249.68 | |
MAGTZUMWQUDWAH-UHFFFAOYSA-N | |
2776411 | |
ClS(=O)(=O)C1=CC=C(S1)C1=CN=CO1 |
C7H4ClNO3S2 | |
MFCD02681961 | |
5-1,3-oxazol-5-yl-2-thiophenesulfonyl chloride, 5-1,3-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-oxazol-5-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-5-oxazolyl, 2-thiophenesulfonyl chloride, 5-5-oxazolyl, chloro 5-1,3-oxazol-5-yl 2-thienyl sulfone, 5-1,3-oxazol-5-yl thiophene-2-sulphonyl chloride, 5-1,3-oxazol-5-yl-2-thiophene sulfonyl chloride, 2-thiophenesulfonyl chloride, 5-5-oxazolyl-9ci | |
5-(1,3-oxazol-5-yl)thiophene-2-sulfonyl chloride |
Specifications
321309-40-4 | |
100.0 | |
C7H4ClNO3S2 | |
5-1,3-oxazol-5-yl-2-thiophenesulfonyl chloride, 5-1,3-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-oxazol-5-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-5-oxazolyl, 2-thiophenesulfonyl chloride, 5-5-oxazolyl, chloro 5-1,3-oxazol-5-yl 2-thienyl sulfone, 5-1,3-oxazol-5-yl thiophene-2-sulphonyl chloride, 5-1,3-oxazol-5-yl-2-thiophene sulfonyl chloride, 2-thiophenesulfonyl chloride, 5-5-oxazolyl-9ci | |
ClS(=O)(=O)C1=CC=C(S1)C1=CN=CO1 | |
249.68 | |
249.7 | |
5-(1, 3-Oxazol-5-yl)-2-thiophenesulfonyl chloride |
96.5 | |
97% | |
MFCD02681961 | |
MAGTZUMWQUDWAH-UHFFFAOYSA-N | |
5-(1,3-oxazol-5-yl)thiophene-2-sulfonyl chloride | |
2776411 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger