missing translation for 'onlineSavingsMsg'
Learn More

5-(1,3-Oxazol-5-yl)-2-thiophenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10542832
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10542832 1 g Amber Glass Bottle 1g
11340577 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10542832 Supplier Thermo Scientific Maybridge Supplier No. CC14203DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 321309-40-4
Molecular Formula C7H4ClNO3S2
Molecular Weight (g/mol) 249.68
MDL Number MFCD02681961
InChI Key MAGTZUMWQUDWAH-UHFFFAOYSA-N
Synonym 5-1,3-oxazol-5-yl-2-thiophenesulfonyl chloride, 5-1,3-oxazol-5-yl thiophene-2-sulfonyl chloride, 5-oxazol-5-yl thiophene-2-sulfonyl chloride, 2-thiophenesulfonylchloride, 5-5-oxazolyl, 2-thiophenesulfonyl chloride, 5-5-oxazolyl, chloro 5-1,3-oxazol-5-yl 2-thienyl sulfone, 5-1,3-oxazol-5-yl thiophene-2-sulphonyl chloride, 5-1,3-oxazol-5-yl-2-thiophene sulfonyl chloride, 2-thiophenesulfonyl chloride, 5-5-oxazolyl-9ci
PubChem CID 2776411
IUPAC Name 5-(1,3-oxazol-5-yl)thiophene-2-sulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=C(S1)C1=CN=CO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 249.7
Percent Purity 97%
Chemical Name or Material 5-(1, 3-Oxazol-5-yl)-2-thiophenesulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.