missing translation for 'onlineSavingsMsg'
Learn More

(4S)-4-Hydroxy-L-isoleucine, TRC

Product Code. 30400076
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
1mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30400076 1 mg 1mg
30408111 10 mg -
30407425 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400076 Supplier Toronto Research Chemicals Supplier No. TRCH9435701MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 55399-93-4
Molecular Formula C6 H13 N O3
Molecular Weight (g/mol) 147.17
Synonym 2-Amino-2,3,5-trideoxy-3-methyl-L-ribonic acid, (2S,3R,4S)-4-Hydroxyisoleucine
IUPAC Name (2R,3R,4R)-2-amino-4-hydroxy-3-methyl-pentanoic acid
SMILES C[C@@H](O)[C@H](C)[C@@H](N)C(=O)O

Specifications

InChI Formula InChI=1S/C6H13NO3/c1-3(4(2)8)5(7)6(9)10/h3-5,8H,7H2,1-2H3,(H,9,10)/t3-,4+,5+/m0/s1
Quantity 1 mg
Formula Weight 147.0895
Shipping Condition Room Temperature
Chemical Name or Material (S)-4-Hydroxy-L-isoleucine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.