CAS RN 55399-93-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(4S)-4-Hydroxy-L-isoleucine, TRC
CAS: 55399-93-4 Molecular Formula: C6 H13 N O3 Molecular Weight (g/mol): 147.17 Synonym: 2-Amino-2,3,5-trideoxy-3-methyl-L-ribonic acid,(2S,3R,4S)-4-Hydroxyisoleucine IUPAC Name: (2R,3R,4R)-2-amino-4-hydroxy-3-methyl-pentanoic acid SMILES: C[C@@H](O)[C@H](C)[C@@H](N)C(=O)O