Learn More
Agilent BioTek 405 TS Microplate Washer
Shop All Agilent BioTek Products

Description
The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. The 405 TS has a unique set of features designed to efficiently wash cell-based assays, microsphere-based assays, and ELISA applications. Programming and operation via the touchscreen interface is intuitive and simple.
The patented Verify technology automatically checks for manifold tube blockages before a wash cycle, and Ultrasonic Advantage automatically cleans any blocked manifold tubes. These functions save time and eliminate frustration while ensuring successful results.
Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.
Spécification
Spécification
| Assays | ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes |
| Certifications/Compliance | CE and TUV marked. RoHS compliant. |
| Connections | USB (2) |
| Bottles | Optional (4 or 10 L) |
| Precision | <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing) |
| Dispense Volume | 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold) |
| Plate Type | 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates |
| Residual Volume | ≤2 μL/well |
| Washing Speed | 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s |
| Soak Time | Programmable in minutes and seconds, up to 60 min. |
| Afficher plus |
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
