missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 405 TS Microplate Washer
GREENER_CHOICE

Product Code. 17247889 Shop All Agilent BioTek Products
Click to view available options
Model:
405TSR
405TSRS
405TSRSQ
405TSRV
405TSRVS
405TSRVSQ
405TSU
405TSUS
405TSUSQ
405TSUV
405TSUVS
405TSUVSQ
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17247889

Brand: Agilent BioTek 405TSUVSQSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications.

The BioTek 405 TS washer has an excellent reputation in life science research for outstanding results in many applications. The 405 TS has a unique set of features designed to efficiently wash cell-based assays, microsphere-based assays, and ELISA applications. Programming and operation via the touchscreen interface is intuitive and simple.

The patented Verify technology automatically checks for manifold tube blockages before a wash cycle, and Ultrasonic Advantage automatically cleans any blocked manifold tubes. These functions save time and eliminate frustration while ensuring successful results.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

TRUSTED_SUSTAINABILITY

Specifications

Assays ELISA, Cell-based assays (model dependent), Magnetic and polystyrene bead, Multiplex assays, Bead-based ELISAs, Filtration-to-waste processes
Certifications/Compliance CE and TUV marked. RoHS compliant.
Connections USB (2)
Bottles Optional (4 or 10 L)
Precision <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing)
Dispense Volume 25 to 3000 μL/well (192-pin manifold); 50 to 3000 μL/well (96-pin manifold)
Plate Type 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Option) Plates
Residual Volume ≤2 μL/well
Washing Speed 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s
Soak Time Programmable in minutes and seconds, up to 60 min.
Type Microplate Washer
Weight (Metric) With internal buffer switching - 16.5 kg; Without - 13.5 kg
Wash Cycles 1 to 250
Wash Volume 25 to 3000 μL/well
Display LED
Flow Rate High to Low Flow; Optimized rates for cell assays
Fluid Delivery Type Internal positive displacement pump
For Use With (Application) ELISA, MSD assays, HCS immune cytochemistry, FLIPR Ca2+ flux, Cell-based assays, Magnetic and polystyrene bead processing (gene expression assays, cytokine assays), ELISPOT assays
Frequency 50/60 Hz
Model 405TSUVSQ
Includes Automated Internal 4-Buffer Switching; Ultrasonic Advantage; Optimized Cell Washing; 96-/384-well microplate washing; Verify technology
Width (English) 14 in.
Width (Metric) 35.6 cm
Height (English) 10 in.
Height (Metric) 25.4 cm
Depth (English) 17 in.
Depth (Metric) 43.2 cm
Automation Interface High Resolution LED Backlit Touch Screen
Weight (English) With internal buffer switching - 36 lb.; Without - 30 lb.
Voltage 100/240 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.