missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek 405LS Microplate Washer
GREENER_CHOICE

Product Code. 17257869 Shop All Agilent BioTek Products
Click to view available options
Model:
405LSHT
405LSHTV
405LSR
405LSRS
405LSRSQ
405LSRV
405LSRVS
405LSRVSQ
405LSU
405LSUS
405LSUSQ
405LSUV
405LSUVS
405LSUVSQ
Includes:
4-Buffer Switching
Microplate Washer
Optimized Cell Washing
Optimized Cell Washing, 4-Buffer Switching
Ultrasonic Advantage
Ultrasonic Advantage, Verify Technology
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17257869

Brand: Agilent BioTek 405LSUSN

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability.

The BioTek 405 LS washer has a well earned reputation as an economical plate washer that offers excellent performance and reliability. Washing protocols range from gentle cell washing to vigorous ELISA workflows. The 405 LS has a simple user interface for creation and execution of wash, dispense, aspirate, and maintenance protocols within the extensive onboard software

For use under computer control, Agilent BioTek LHC software used with the 405 LS is available in standard and 21 CFR Part 11 compliant versions. The optional Agilent BioTek BioStack is available for walk-away automation of up to 50 microplates.

 

Must be purchased together with required dispense/waste system accessory for use. Please contact your Fisher Sales Representative for more information.

Order Info

Required for operation: BT1170530, Complete Dispense/Waste System, 4L bottles. Must order separately.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE and TUV marked. RoHS compliant.
Bottles Optional (4 or 10 L)
Precision <3% CV: 300 μL/well (96-well washing); <4% CV: 80 μL/well (384-well washing)
Dispense Volume 25 to 3000 μL/well
Plate Type 96- and 384-well, Low Profile and Standard, Solid and Filter Bottom (Pore Sizes 0.45 to 1.2 μm) Plates
Residual Volume <2 μL/well (96- and 384-well plates), 96-tube manifold for 96 wells, 192-tube for 384 wells
Washing Speed 96-wells: 300 μL/well, 3 cycles: <30 s; 384-wells: 100 μL/well, 3 cycles: <80 s; 384-wells: 400 μL/well, 1 cycle: <20 s
Soak Time Programmable in minutes and seconds, up to 60 min.
Type Microplate Washer
Weight (Metric) With internal buffer switching - 16.5 kg; Without - 13.5 kg
Wash Cycles 1 to 250
Flow Rate High to Low Flow; Optimized rates for cell assays
For Use With (Application) 96/384-well Plate Washing
Humidity 10 to 85%, noncondensing
Frequency 50/60 Hz
Model 405LSU
Increments 1 μL
Includes Optimized Cell Washing
Width (English) 14 in.
Width (Metric) 35.6 cm
Height (English) 10 in.
Height (Metric) 25.4 cm
Depth (English) 17 in.
Depth (Metric) 43.2 cm
Weight (English) With internal buffer switching - 36 lb.; Without - 30 lb.
Voltage 100/240 V
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.