missing translation for 'onlineSavingsMsg'
Learn More

4-(Phenylamino]-1-benzyl-4-piperidinecarboxylic Acid, TRC

Product Code. 30428605
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30428605 500 mg
30421051 250 mg
30418522 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30428605 Supplier Toronto Research Chemicals Supplier No. TRCP307490500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85098-64-2
Molecular Formula C19H22N2O2
Molecular Weight (g/mol) 310.39
Synonym 1-Benzyl-4-(phenylamino)piperidine-4-carboxylic Acid, 4-Anilino-1-benzylpiperidine-4-carboxylic Acid, NSC 73748
IUPAC Name 4-anilino-1-benzylpiperidine-4-carboxylic acid
SMILES OC(=O)C1(CCN(Cc2ccccc2)CC1)Nc3ccccc3

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C19H22N2O2/c22-18(23)19(20-17-9-5-2-6-10-17)11-13-21(14-12-19)15-16-7-3-1-4-8-16/h1-10,20H,11-15H2,(H,22,23)
Formula Weight 310.1681
Chemical Name or Material 4-(Phenylamino)-1-benzyl-4-piperidinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.