CAS RN 85098-64-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(Phenylamino]-1-benzyl-4-piperidinecarboxylic Acid, TRC
CAS: 85098-64-2 Molecular Formula: C19H22N2O2 Molecular Weight (g/mol): 310.39 Synonym: 1-Benzyl-4-(phenylamino)piperidine-4-carboxylic Acid,4-Anilino-1-benzylpiperidine-4-carboxylic Acid,NSC 73748 IUPAC Name: 4-anilino-1-benzylpiperidine-4-carboxylic acid SMILES: OC(=O)C1(CCN(Cc2ccccc2)CC1)Nc3ccccc3