missing translation for 'onlineSavingsMsg'
Learn More

4-Octylbenzenesulfonic Acid Sodium Salt, TRC

Product Code. 30423931
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30423931 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30423931 Supplier Toronto Research Chemicals Supplier No. TRCO2314152.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6149-03-7
Molecular Formula C14H21NaO3S
Molecular Weight (g/mol) 292.37
IUPAC Name sodium;4-octylbenzenesulfonate
SMILES [Na+].CCCCCCCCc1ccc(cc1)S(=O)(=O)[O-]

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C14H22O3S.Na/c1-2-3-4-5-6-7-8-13-9-11-14(12-10-13)18(15,16)17;/h9-12H,2-8H2,1H3,(H,15,16,17);/q;+1/p-1
Formula Weight 292.1109
Chemical Name or Material 4-Octylbenzenesulfonic Acid Sodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.