accessibility menu, dialog, popup

UserName

NaH3CSOOO

CAS RN 6149-03-7

IUPAC Name: sodium 4-octylbenzene-1-sulfonate
Synonyms: sodium 4-octylbenzenesulfonate
Molecular Weight (g/mol): 292.37
Molecular Formula: C14H21NaO3S
InChi Key: ASJUTVUXOMPOQH-UHFFFAOYSA-M
SMILES: [Na+].CCCCCCCCC1=CC=C(C=C1)S([O-])(=O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results