missing translation for 'onlineSavingsMsg'
Learn More

4-O-Alpha-D-Glucopyranosylmoranoline, TRC

Product Code. 30397823
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30397823 1 mg
30405463 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30397823 Supplier Toronto Research Chemicals Supplier No. TRCG4500001MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80312-32-9
Molecular Formula C12 H23 N O9
Molecular Weight (g/mol) 325.31
IUPAC Name (2S,3R,4S,5S,6R)-2-[(2R,3R,4R,5S)-4,5-dihydroxy-2-(hydroxymethyl)piperidin-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES OC[C@H]1NC[C@H](O)[C@@H](O)[C@@H]1O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C12H23NO9/c14-2-4-11(7(17)5(16)1-13-4)22-12-10(20)9(19)8(18)6(3-15)21-12/h4-20H,1-3H2/t4-,5+,6-,7-,8-,9+,10-,11-,12-/m1/s1
Formula Weight 325.1373
Chemical Name or Material 4-O-a-D-Glucopyranosylmoranoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.