CAS RN 80312-32-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-O-Alpha-D-Glucopyranosylmoranoline, TRC
CAS: 80312-32-9 Molecular Formula: C12 H23 N O9 Molecular Weight (g/mol): 325.31 IUPAC Name: (2S,3R,4S,5S,6R)-2-[(2R,3R,4R,5S)-4,5-dihydroxy-2-(hydroxymethyl)piperidin-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: OC[C@H]1NC[C@H](O)[C@@H](O)[C@@H]1O[C@H]2O[C@H](CO)[C@@H](O)[C@H](O)[C@H]2O